Learn More
Sino Biological Recombinant Human SPINK2 Protein (hFc Tag)
Supplier:  Sino Biological 13636H02H100
Encompass_Preferred
A DNA sequence encoding the human SPINK2 (NP_066937.1) (Met1-Cys84) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8718