Learn More
Sino Biological Recombinant Human BMF Protein (His Tag)
Supplier:  Sino Biological 13639H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human BMF (Q96LC9-1) (Met1-Arg184) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8720