Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches

Catalog No. 13641276
Change view
Click to view available options
Quantity:
10 x 15 mL
25 x 15 mL
pH:
4.01
7
10.01
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity pH
13-641-276 25 x 15 mL 10.01
13-641-270 10 x 15 mL 4.01
13-641-274 25 x 15 mL 4.01
13-641-271 10 x 15 mL 7
13-641-275 25 x 15 mL 7
13-641-272 10 x 15 mL 10.01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 13-641-276 Supplier Thermo Scientific™ Supplier No. 911025WA
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.

Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs

Note:

The following part numbers have been changed due to a recent system upgrade.

Old Part Number: 910410

New Part Number: 910410-WA

Old Part Number: 911025

New Part Number: 911025-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Buffer, individual use pouches
Boiling Point 100°C
Color Light Blue
Description pH 10.01 Buffer
For Use With (Application) pH calibration
For Use With (Equipment) pH Meter and Electrode, single-use
pH 10.01
Physical Form Liquid
Quantity 25 x 15 mL
Solution Type Buffer
Formulation Water, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate
Packaging Pouch
Traceability to NIST Yes
Solubility Information Soluble
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.