Learn More
Sino Biological Recombinant Human NCKIPSD Protein (GST Tag)
Supplier:  Sino Biological 13642H09E100
Encompass_Preferred
A DNA sequence encoding the mature form of human NCKIPSD (Q9NZQ3-3) (Met1-Thr244) was expressed with the GST tag at the N-terminus.
Catalog No. 50-161-8722