Learn More
Sino Biological Recombinant Human CYTL1 Protein (mFc Tag)
Supplier:  Sino Biological 13647H05H100
Encompass_Preferred
A DNA sequence encoding the human CYTL1 (NP_061129.1) (Met1-Arg136) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8731