Learn More
Sino Biological Recombinant Human Serglycin/SRGN Protein (His & Myc Tag), HPLC-verified
Supplier:  Sino Biological 13648H33H100
Encompass_Preferred
A DNA sequence encoding the human SRGN (AAA60179.1) (Met1-Leu158) was fused with the C-terminal polyhistidine-tagged myc at the C-terminus.
Catalog No. 50-161-8735