Learn More
Sino Biological Recombinant Human PRAK/MAPKAPK5 Protein (His & GST Tag)
Supplier:  Sino Biological 13655H20B50
Encompass_Preferred
A DNA sequence encoding the human MAPKAPK5 isoform 1 (NP_003659.2) (Met 1-Gln 471) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8748