Learn More
Sino Biological Recombinant Sus scrofa (Pig) CD63 Protein (hFc Tag)
Supplier:  Sino Biological 13657W02H100
Encompass_Preferred
A DNA sequence encoding the Sus scrofa (Pig) CD63 (XP_003355519.1) (Arg 108-Phe 204) was fused with the human IgG1 Fc region at the N-terminus.
Catalog No. 50-161-8753