Learn More
Sino Biological Recombinant Human GNGT1 Protein (His Tag)
Supplier:  Sino Biological 13658H07E100
Encompass_Preferred
A DNA sequence encoding the mature foem of human GNGT1 (P63211) (Pro 2-Cys 71) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8755