Learn More
Sino Biological Recombinant Human TMUB2 Protein (His Tag)
Supplier:  Sino Biological 13660H07H100
Encompass_Preferred
A DNA sequence encoding the human TMUB2 (NP_803190.2) (Tyr37-Ser245) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-8759