Learn More
Sino Biological Recombinant Human BPHL Protein (His Tag)
Supplier:  Sino Biological 13675H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human BPHL (Q86WA6-1) (Ser38-Gln291) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8769