Learn More
Sino Biological Recombinant Human NAGA Protein (His Tag)
Supplier:  Sino Biological 13686H08H50
Encompass_Preferred
A DNA sequence encoding the human NAGA (NP_000253.1) (Met1-Gln411) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8774