Learn More
Sino Biological Recombinant Human PLRP2 Protein (His Tag)
Supplier:  Sino Biological 13687H08B100
Encompass_Preferred
A DNA sequence encoding the human PNLIPRP2 (P54317) (Met 1-Cys469) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-8775