Learn More
Sino Biological Recombinant Human APLP1 Protein (His Tag)
Supplier:  Sino Biological 13716H08H100
Encompass_Preferred
A DNA sequence encoding the human APLP1 (P51693-1) (Met1-Glu580) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8785