Learn More
Sino Biological Recombinant Human TMED4 Protein (mFc Tag)
Supplier:  Sino Biological 13729H05H100
Encompass_Preferred
A DNA sequence encoding the human TMED4 (NP_872353.2) (Met1-Arg194) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8791