Learn More
Sino Biological Recombinant Human BOLA1 Protein (His Tag)
Supplier:  Sino Biological 13743H07E100
Encompass_Preferred
A DNA sequence encoding the human BOLA1 (Q9Y3E2) (Met1-Pro137) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8797