Learn More
Sino Biological Recombinant Human PILR-beta Protein (His Tag)
Supplier:  Sino Biological 13746H07H200
Encompass_Preferred
A DNA sequence encoding the human PILRB (NP_839956.1) (Gln20-Ala189) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8801