Learn More
Sino Biological Recombinant Human CTRL Protein (His Tag)
Supplier:  Sino Biological 13748H08H100
Encompass_Preferred
A DNA sequence encoding the human CTRL (P40313) (Met1-Asn264) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8803