Learn More
Sino Biological Recombinant Human GALNT2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13764H07H50
Encompass_Preferred
A DNA sequence encoding the human GALNT2 (NP_004472.1) (Lys52-Gln571) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-8818