Learn More
Sino Biological Recombinant Human CNPY2 Protein (His Tag)
Supplier:  Sino Biological 13771H08H100
Encompass_Preferred
A DNA sequence encoding the human CNPY2 (Q9Y2B0-1) (Met1-Ser178) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8821