Learn More
Sino Biological Recombinant Human PILR-alpha/FDF03 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 13791H02H100
Encompass_Preferred
A DNA sequence encoding the human PILRA (AAH17812.1)(Met1-Asp226) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8827