Learn More
Sino Biological Recombinant Human DNAJC30 Protein (His Tag)
Supplier:  Sino Biological 13796H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human DNAJC30 (Q96LL9) (Ser39-Gly124) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8835