Learn More
Sino Biological Recombinant Human DPEP2 Protein (hFc Tag)
Supplier:  Sino Biological 13806H02H100
Encompass_Preferred
A DNA sequence encoding the human DPEP2 (AAH24021.1) (Met1-Ser376) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8841