Learn More
Sino Biological Recombinant Human NRN1L Protein (mFc Tag)
Supplier:  Sino Biological 13815H05H100
Encompass_Preferred
A DNA sequence encoding the human NRN1L (Q496H8) (Met1-Thr138) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8847