Learn More
Sino Biological Recombinant Human LMAN2L Protein (His Tag)
Supplier:  Sino Biological 13822H08H50
Encompass_Preferred
A DNA sequence encoding the human LMAN2L (Q9H0V9-1) (Met1-Ala304) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8856