Learn More
Sino Biological Recombinant Human IL-23 Receptor Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 13840H02H100
Encompass_Preferred
A DNA sequence encoding the human IL23R (AAM44229.1) (Met1-Asp353) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8859