Learn More
Sino Biological Recombinant Human UBASH3A Protein (aa 354-623, His Tag)
Supplier:  Sino Biological 13847H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human UBASH3A (AAH69511.1) (Ala354-Asn623) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8861