Learn More
Sino Biological Recombinant Human Complement C7 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13848H08H100
Encompass_Preferred
A DNA sequence encoding the human C7 (P10643) (Met1-Gln843) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-8865