Learn More
Sino Biological Recombinant Human SLITL2/VASN Protein (His Tag)
Supplier:  Sino Biological 13854H08H100
Encompass_Preferred
A DNA sequence encoding the human VASN (Q6EMK4) (Met1-Pro575) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8867