Learn More
Sino Biological Recombinant Human SLC27A4/FATP4 Protein (His Tag)
Supplier:  Sino Biological 13861H07H100
Encompass_Preferred
A DNA sequence encoding the human SLC27A4 (NP_005085.2) (Gln483-Leu643) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-161-8875