Learn More
Sino Biological Recombinant Human RFK Protein (His Tag)
Supplier:  Sino Biological 13881H07E50
Encompass_Preferred
A DNA sequence encoding the mature form of human RFK (Q969G6) (Met1-His155) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8883