Learn More
Sino Biological Recombinant Human FUCA1 Protein (His Tag)
Supplier:  Sino Biological 13893H08H20
Encompass_Preferred
A DNA sequence encoding the human FUCA1 (P04066) (Gln32-Lys466) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8885