Learn More
Sino Biological Recombinant Human GPHA2 Protein (His Tag)
Supplier:  Sino Biological 13894H08H100
Encompass_Preferred
A DNA sequence encoding the human GPHA2 (Q96T91) (Met1-Tyr129) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8888