Learn More
Sino Biological Recombinant Human APOL1 Protein (His Tag)
Supplier:  Sino Biological 13910H08B50
Encompass_Preferred
A DNA sequence encoding the human APOL1 (Q2KHQ6) (Met1-Leu398) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-8905