Learn More
Sino Biological Recombinant Human MMP19 Protein
Supplier:  Sino Biological 13912HNAE20
Encompass_Preferred
A DNA sequence encoding the catalytic domain of human MMP19 (Q99542-1) (Leu101-Gly256) was expressed, with a N-terminal Met.
Catalog No. 50-161-8909