Learn More
Sino Biological Recombinant Human ABHD14B Protein (His Tag)
Supplier:  Sino Biological 13913H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human ABHD14B (Q96IU4-1) (Met1-Gln210) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8910