Learn More
Sino Biological Recombinant Human HMGN3 Protein (His Tag)
Supplier:  Sino Biological 13914H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human HMGN3 (Q15651-2) (Met1-Asn77) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8912