Learn More
Sino Biological Recombinant Human BPIFA2 Protein (hFc Tag)
Supplier:  Sino Biological 13915H02H100
Encompass_Preferred
A DNA sequence encoding the human C20orf70 (Q96DR5) (Met1-Ile249) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8914