Learn More
Sino Biological Recombinant Human BPIFA2 Protein (His Tag)
Supplier:  Sino Biological 13915H08H100
Encompass_Preferred
A DNA sequence encoding the human C20orf70 (Q96DR5) (Met1-Ile249) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8917