Learn More
Sino Biological Recombinant Human BLOC1S2 Protein (GST Tag)
Supplier:  Sino Biological 13916H09E100
Encompass_Preferred
A DNA sequence encoding the human BLOC1S2 (Q6QNY1-2) (Met1-Arg99) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8919