Learn More
Sino Biological Recombinant Human Tubulin cofactor A Protein
Supplier:  Sino Biological 13918HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human TBCA (O75347) (Met1-Ala108) was expressed, with a N-terminal Met.
Catalog No. 50-161-8923