Learn More
Sino Biological Recombinant Human Parvin alpha/PARVA Protein (GST Tag)
Supplier:  Sino Biological 13919H09E100
Encompass_Preferred
A DNA sequence encoding the mature form of human PARVA (Q9NVD7-1) (Met1-Glu372) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8925