Learn More
Sino Biological Recombinant Human SAE1 Protein (His & GST Tag)
Supplier:  Sino Biological 13921H20B100
Encompass_Preferred
A DNA sequence encoding the human SAE1 (Q9UBE0) (Met1-Lys346) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8929