Learn More
Sino Biological Recombinant Human CKMT1A Protein (His Tag)
Supplier:  Sino Biological 13924H07B150
Encompass_Preferred
A DNA sequence encoding human CKMT1A (P12532-1) (Ala40-His417) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8937