Learn More
Sino Biological Recombinant Human IP6K1/IHPK1 Protein (His & GST Tag)
Supplier:  Sino Biological 13927H20B50
Encompass_Preferred
A DNA sequence encoding the human IHPK1 (Q92551-1) (Met1-Gln441) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8944