Learn More
Sino Biological Recombinant Human EIF5 Protein (GST Tag)
Supplier:  Sino Biological 13932H09E100
Encompass_Preferred
A DNA sequence encoding the mature form of human EIF5 (P55010) (Met1-Asp150) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8951