Learn More
Sino Biological Recombinant Human PLBL2 / PLBD2 Protein (His Tag)
Supplier:  Sino Biological 13947H08H100
Encompass_Preferred
A DNA sequence encoding the human PLBD2 (Q8NHP8) (Met1-Asp589) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8955