Learn More
Cole-Parmer Pyrex 1395-50 Brand 1395 Media Storage Bottle w/ Screw Cap, 50 mL
Catalog No. NC0052052 Shop All Cole-Parmer Products
Encompass_Preferred
Bottles have permanent white enamel graduations and marking spots. Caution: Do not tighten caps immediately after autoclaving.
This item is not returnable. View return policy
Catalog No. NC0052052 Supplier Cole-Parmer Supplier No. 3451421
Bottles have permanent white enamel graduations and marking spots. Caution: Do not tighten caps immediately after autoclaving.