Learn More
Sino Biological Recombinant Human B3GNT6 Protein (aa 44-384, His Tag)
Supplier:  Sino Biological 13951H07B50
Encompass_Preferred
A DNA sequence encoding the human B3GNT6 (NP_619651.3)(Gln44-Ser384) was fused with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8958