Learn More
Sino Biological Recombinant Human LRP11 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13952H08H100
Encompass_Preferred
A DNA sequence encoding the human LRP11 (Q86VZ4-1) (Met1-Gly450) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-8959